Assay Details
| Gene Symbol: il1rapl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077576.1 | NP_001071044.1 | 9 - 10 | 1208 | 77 | |
| XM_005174698.4 | XP_005174755.1 | 7 - 8 | 77 | |||
| XM_009304759.3 | XP_009303034.1 | 7 - 8 | 77 | |||
| XM_009304760.3 | XP_009303035.1 | 7 - 8 | 77 | |||
| XM_017357710.2 | XP_017213199.1 | 7 - 8 | 77 | |||
| XM_021478747.1 | XP_021334422.1 | 9 - 10 | 77 | |||
| XM_021478748.1 | XP_021334423.1 | 7 - 8 | 77 | |||
| GenBank mRNA | AB449367.1 | - | 6 - 7 | 77 | ||
| BC124594.1 | - | 9 - 10 | 1208 | 77 | ||
More Information
Set Membership: |
|

il1rapl1a
Genomic Map