Assay Details
| Gene Symbol: peli1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077601.2 | NP_001071069.2 | 2 - 3 | 653 | 66 | |
| GenBank mRNA | BC134833.1 | - | 2 - 3 | 653 | 66 | |
More Information
Set Membership: |
|

peli1b
Genomic Map