Assay Details
| Gene Symbol: il20ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079676.2 | NP_001073144.2 | 5 - 6 | 847 | 102 | |
| XM_005160336.4 | XP_005160393.1 | 5 - 6 | 853 | 102 | ||
| GenBank mRNA | AM084416.2 | - | 5 - 6 | 797 | 102 | |
| EF014958.1 | - | 5 - 6 | 839 | 102 | ||
More Information
Set Membership: |
|

il20ra
Genomic Map