Assay Details
| Gene Symbol: rtn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004555.2 | NP_001004555.2 | 4 - 5 | 580 | 62 | |
| NM_001079912.2 | NP_001073381.1 | 4 - 5 | 1099 | 62 | ||
| NM_001201446.1 | NP_001188375.1 | 4 - 5 | 501 | 62 | ||
| GenBank mRNA | AY555039.1 | - | 4 - 5 | 1026 | 62 | |
| AY555040.1 | - | 5 - 6 | 821 | 62 | ||
| AY555041.1 | - | 5 - 6 | 775 | 62 | ||
| AY555042.1 | - | 4 - 5 | 579 | 62 | ||
| AY555043.1 | - | 4 - 5 | 481 | 62 | ||
| BC081614.1 | - | 4 - 5 | 567 | 62 | ||
| EF061088.1 | - | 4 - 5 | 1090 | 62 | ||
| EF061089.1 | - | 4 - 5 | 578 | 62 | ||
| EF061090.1 | - | 4 - 5 | 501 | 62 | ||
More Information
Set Membership: |
|

rtn4a
Genomic Map