Assay Details
| Gene Symbol: retreg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080014.1 | NP_001073483.1 | 2 - 3 | 396 | 68 | |
| XM_005160291.3 | XP_005160348.1 | 2 - 3 | 496 | 68 | ||
| GenBank mRNA | BC128884.1 | - | 2 - 3 | 396 | 68 | |
More Information
Set Membership: |
|

retreg2
Genomic Map