Assay Details
Gene Symbol: arrdc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080029.1 | NP_001073498.1 | 2 - 3 | 547 | 97 | |
GenBank mRNA | BC117667.1 | - | 2 - 3 | 547 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
