Assay Details
| Gene Symbol: rad21l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080050.1 | NP_001073519.1 | 6 - 7 | 637 | 67 | |
| XM_021478297.1 | XP_021333972.1 | 5 - 6 | 668 | 67 | ||
| GenBank mRNA | BC125933.1 | - | 6 - 7 | 637 | 67 | |
More Information
Set Membership: |
|

rad21l1
Genomic Map