Assay Details
| Gene Symbol: rad21l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080050.1 | NP_001073519.1 | 8 - 9 | 864 | 64 | |
| XM_021478297.1 | XP_021333972.1 | 7 - 8 | 895 | 64 | ||
| GenBank mRNA | BC125933.1 | - | 8 - 9 | 864 | 64 | |
More Information
Set Membership: |
|

rad21l1
Genomic Map