Assay Details
| Gene Symbol: mxra8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080582.1 | NP_001074051.1 | 2 - 3 | 216 | 81 | |
| GenBank mRNA | BC129223.1 | - | 2 - 3 | 216 | 81 | |
| CU638765.1 | - | 2 - 3 | 216 | 81 | ||
More Information
Set Membership: |
|

mxra8a
Genomic Map