Assay Details
| Gene Symbol: mxra8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080582.1 | NP_001074051.1 | 6 - 7 | 1243 | 79 | |
| XM_009306033.3 | XP_009304308.1 | 7 - 8 | 1346 | 79 | ||
| XM_021479639.1 | XP_021335314.1 | 7 - 8 | 1347 | 79 | ||
| GenBank mRNA | BC129223.1 | - | 6 - 7 | 1243 | 79 | |
| CU638765.1 | - | 6 - 7 | 1243 | 79 | ||
More Information
Set Membership: |
|

mxra8a
Genomic Map