Assay Details
| Gene Symbol: mxra8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080582.1 | NP_001074051.1 | 7 - 8 | 1289 | 103 | |
| XM_009306033.3 | XP_009304308.1 | 8 - 9 | 1392 | 103 | ||
| GenBank mRNA | BC129223.1 | - | 7 - 8 | 1289 | 103 | |
| CU638765.1 | - | 7 - 8 | 1289 | 103 | ||
More Information
Set Membership: |
|

mxra8a
Genomic Map