Assay Details
| Gene Symbol: rorca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082819.1 | NP_001076288.1 | 3 - 4 | 693 | 112 | |
| GenBank mRNA | DQ017627.1 | - | 3 - 4 | 693 | 112 | |
| EF107094.1 | - | 3 - 4 | 693 | 112 | ||
More Information
Set Membership: |
|

rorca
Genomic Map