Assay Details
| Gene Symbol: pld6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089414.1 | NP_001082883.1 | 2 - 3 | 277 | 115 | |
| GenBank mRNA | BC122189.1 | - | 2 - 3 | 115 | ||
| BC134035.1 | - | 2 - 3 | 277 | 115 | ||
More Information
Set Membership: |
|

pld6
Genomic Map