Assay Details
Gene Symbol: LOC558044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099989.1 | NP_001093459.1 | 32 - 33 | 5954 | 104 | |
XM_005165320.4 | XP_005165377.1 | 32 - 33 | 6345 | 104 | ||
XM_005165321.4 | XP_005165378.1 | 31 - 32 | 6343 | 104 | ||
XM_021476187.1 | XP_021331862.1 | 31 - 32 | 5846 | 104 | ||
XM_021476188.1 | XP_021331863.1 | 31 - 32 | 5846 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
