Assay Details
| Gene Symbol: dkk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131003.1 | NP_571078.1 | 1 - 2 | 197 | 66 | |
| GenBank mRNA | AF116852.1 | - | 1 - 2 | 197 | 66 | |
| KM655708.1 | - | 1 - 2 | 192 | 66 | ||
More Information
Set Membership: |
|

dkk1b
Genomic Map