Assay Details
| Gene Symbol: dkk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131003.1 | NP_571078.1 | 3 - 4 | 482 | 95 | |
| XM_005172963.3 | XP_005173020.1 | 3 - 4 | 558 | 95 | ||
| GenBank mRNA | AB023488.1 | - | 3 - 4 | 534 | 95 | |
| AF116852.1 | - | 3 - 4 | 482 | 95 | ||
| BC124128.1 | - | 3 - 4 | 526 | 95 | ||
| BC165086.1 | - | 3 - 4 | 508 | 95 | ||
| JN966755.1 | - | 3 - 4 | 518 | 95 | ||
| KM655708.1 | - | 3 - 4 | 477 | 95 | ||
More Information
Set Membership: |
|

dkk1b
Genomic Map