Assay Details
| Gene Symbol: sema3aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131060.1 | NP_571135.1 | 11 - 12 | 1717 | 67 | |
| GenBank mRNA | AF086761.1 | - | 11 - 12 | 1717 | 67 | |
| BC162381.1 | - | 11 - 12 | 1430 | 67 | ||
More Information
Set Membership: |
|

sema3aa
Genomic Map