Assay Details
| Gene Symbol: tpma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131105.2 | NP_571180.1 | 6 - 7 | 690 | 75 | |
| GenBank mRNA | AF180892.1 | - | 6 - 7 | 701 | 75 | |
| BC062870.1 | - | 6 - 7 | 690 | 75 | ||
More Information
Set Membership: |
|

tpma
Genomic Map