Assay Details
Gene Symbol: pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131113.1 | NP_571188.1 | 7 - 8 | 1109 | 61 | |
XM_005162296.4 | XP_005162353.1 | 6 - 7 | 1235 | 61 | ||
XM_017353502.2 | XP_017208991.1 | 7 - 8 | 1315 | 61 | ||
GenBank mRNA | BC162134.1 | - | 7 - 8 | 951 | 61 | |
BC162146.1 | - | 7 - 8 | 951 | 61 | ||
X68432.1 | - | 1109 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
