Assay Details
| Gene Symbol: fzd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131134.2 | NP_571209.2 | 1 | 1503 | 75 | |
| GenBank mRNA | AF117387.1 | - | 1 | 75 | ||
| BC163118.1 | - | 1 | 1503 | 75 | ||
| BC163120.1 | - | 1 | 1503 | 75 | ||
More Information
Set Membership: |
|

fzd5
Genomic Map