Assay Details
| Gene Symbol: fzd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131140.1 | NP_571215.1 | 1 | 1651 | 88 | |
| GenBank mRNA | AY033592.1 | - | 1 | 1651 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fzd2| Gene Symbol: fzd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131140.1 | NP_571215.1 | 1 | 1651 | 88 | |
| GenBank mRNA | AY033592.1 | - | 1 | 1651 | 88 | |
Set Membership: |
|