Assay Details
| Gene Symbol: rxrab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131153.1 | NP_571228.1 | 5 - 6 | 720 | 78 | |
| XM_009301955.3 | XP_009300230.1 | 7 - 8 | 1201 | 78 | ||
| GenBank mRNA | U29894.1 | - | 5 - 6 | 720 | 78 | |
More Information
Set Membership: |
|

rxrab
Genomic Map