Assay Details
| Gene Symbol: atp1b3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131221.2 | NP_571296.2 | 1 - 2 | 347 | 85 | |
| XM_021478725.1 | XP_021334400.1 | 1 - 2 | 350 | 85 | ||
| XM_021478732.1 | XP_021334407.1 | 1 - 2 | 348 | 85 | ||
| GenBank mRNA | AF469651.1 | - | 1 - 2 | 205 | 85 | |
| EH580086.1 | - | 1 - 2 | 377 | 85 | ||
| X89722.1 | - | 265 | 85 | |||
More Information
Set Membership: |
|

atp1b3a
Genomic Map