Assay Details
| Gene Symbol: atp1b3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131221.2 | NP_571296.2 | 3 - 4 | 568 | 139 | |
| XM_021478725.1 | XP_021334400.1 | 3 - 4 | 571 | 139 | ||
| XM_021478732.1 | XP_021334407.1 | 3 - 4 | 569 | 139 | ||
| GenBank mRNA | AF469651.1 | - | 3 - 4 | 426 | 139 | |
| BC059426.1 | - | 3 - 4 | 540 | 139 | ||
| BC065944.1 | - | 3 - 4 | 356 | 139 | ||
| EH580086.1 | - | 3 - 4 | 598 | 139 | ||
| X89722.1 | - | 486 | 139 | |||
More Information
Set Membership: |
|

atp1b3a
Genomic Map