Assay Details
| Gene Symbol: fgf8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131281.2 | NP_571356.2 | 4 - 5 | 817 | 104 | |
| GenBank mRNA | AF034264.1 | - | 4 - 5 | 359 | 104 | |
| AF051365.1 | - | 4 - 5 | 104 | |||
| BC086718.1 | - | 4 - 5 | 817 | 104 | ||
| BC165018.1 | - | 4 - 5 | 385 | 104 | ||
| KM655579.1 | - | 4 - 5 | 351 | 104 | ||
More Information
Set Membership: |
|

fgf8a
Genomic Map