Assay Details
| Gene Symbol: tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131432.2 | NP_571507.2 | 10 - 11 | 1500 | 67 | |
| GenBank mRNA | AF021344.1 | - | 10 - 11 | 1533 | 67 | |
| BC162462.1 | - | 10 - 11 | 1506 | 67 | ||
| CU458897.1 | - | 10 - 11 | 1304 | 67 | ||
More Information
Set Membership: |
|

tyro3
Genomic Map