Assay Details
| Gene Symbol: prph2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131566.1 | NP_571641.1 | 1 - 2 | 837 | 70 | |
| GenBank mRNA | BC081641.1 | - | 1 - 2 | 837 | 70 | |
More Information
Set Membership: |
|

prph2a
Genomic Map