Assay Details
| Gene Symbol: dap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131573.2 | NP_571648.1 | 1 - 2 | 112 | 77 | |
| XM_017357715.2 | XP_017213204.1 | 1 - 2 | 151 | 77 | ||
| GenBank mRNA | AF231128.1 | - | 1 - 2 | 58 | 77 | |
| BC091443.1 | - | 1 - 2 | 108 | 77 | ||
| BC115168.1 | - | 1 - 2 | 112 | 77 | ||
| Gene Symbol: LOC101885164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017357859.2 | XP_017213348.1 | 174 | 77 | ||
More Information
Set Membership: |
|

dap1b
Genomic Map