Assay Details
| Gene Symbol: mibp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131693.2 | NP_571768.2 | 2 - 3 | 293 | 60 | |
| XM_017353192.2 | XP_017208681.1 | 2 - 3 | 352 | 60 | ||
| GenBank mRNA | AF195851.1 | - | 2 - 3 | 240 | 60 | |
| BC050155.1 | - | 2 - 3 | 249 | 60 | ||
| BC152226.1 | - | 2 - 3 | 248 | 60 | ||
| EH550773.1 | - | 2 - 3 | 332 | 60 | ||
More Information
Set Membership: |
|

mibp
Genomic Map