Assay Details
| Gene Symbol: mibp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131693.2 | NP_571768.2 | 3 - 4 | 342 | 65 | |
| XM_017353192.2 | XP_017208681.1 | 3 - 4 | 401 | 65 | ||
| GenBank mRNA | AF195851.1 | - | 3 - 4 | 289 | 65 | |
| BC050155.1 | - | 3 - 4 | 298 | 65 | ||
| EH550773.1 | - | 3 - 4 | 381 | 65 | ||
More Information
Set Membership: |
|

mibp
Genomic Map