Assay Details
| Gene Symbol: mibp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131693.2 | NP_571768.2 | 5 - 6 | 590 | 102 | |
| NR_102288.1 | 4 - 5 | 544 | 102 | |||
| XM_017353192.2 | XP_017208681.1 | 5 - 6 | 649 | 102 | ||
| XM_017353193.2 | XP_017208682.1 | 4 - 5 | 593 | 102 | ||
| GenBank mRNA | AF195851.1 | - | 5 - 6 | 538 | 103 | |
| BC050155.1 | - | 5 - 6 | 546 | 102 | ||
| BC152226.1 | - | 5 - 6 | 545 | 102 | ||
| EH550773.1 | - | 5 - 6 | 629 | 102 | ||
More Information
Set Membership: |
|

mibp
Genomic Map