Assay Details
| Gene Symbol: mibp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131693.2 | NP_571768.2 | 7 - 8 | 775 | 64 | |
| NR_102288.1 | 6 - 7 | 729 | 64 | |||
| GenBank mRNA | AF195851.1 | - | 7 - 8 | 723 | 64 | |
| BC050155.1 | - | 7 - 8 | 731 | 64 | ||
| BC152226.1 | - | 7 - 8 | 730 | 64 | ||
| EH550773.1 | - | 7 - 8 | 814 | 64 | ||
More Information
Set Membership: |
|

mibp
Genomic Map