Assay Details
| Gene Symbol: rpa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131711.2 | NP_571786.2 | 5 - 6 | 526 | 61 | |
| GenBank mRNA | AY007304.1 | - | 5 - 6 | 458 | 61 | |
| BC095005.1 | - | 5 - 6 | 526 | 61 | ||
| BC164047.1 | - | 4 - 5 | 426 | 61 | ||
More Information
Set Membership: |
|

rpa2
Genomic Map