Assay Details
| Gene Symbol: mdkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131716.1 | NP_571791.1 | 2 - 3 | 129 | 91 | |
| GenBank mRNA | AY008836.1 | - | 2 - 3 | 129 | 91 | |
| KM655598.1 | - | 1 - 2 | 81 | 91 | ||
More Information
Set Membership: |
|

mdkb
Genomic Map