Assay Details
| Gene Symbol: igf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131825.2 | NP_571900.1 | 3 - 4 | 592 | 66 | |
| GenBank mRNA | AF268051.1 | - | 3 - 4 | 511 | 66 | |
| AF314545.1 | - | 3 - 4 | 493 | 66 | ||
| BC114262.1 | - | 3 - 4 | 592 | 66 | ||
More Information
Set Membership: |
|

igf1
Genomic Map