Assay Details
| Gene Symbol: tfe3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131848.2 | NP_571923.2 | 1 - 2 | 500 | 83 | |
| GenBank mRNA | AF362684.1 | - | 1 - 2 | 331 | 83 | |
| BC092697.1 | - | 1 - 2 | 500 | 83 | ||
| BC165184.1 | - | 1 - 2 | 165 | 83 | ||
More Information
Set Membership: |
|

tfe3a
Genomic Map