Assay Details
| Gene Symbol: tfe3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131848.2 | NP_571923.2 | 7 - 8 | 1340 | 81 | |
| XM_005166704.4 | XP_005166761.1 | 7 - 8 | 1104 | 81 | ||
| XM_005166705.4 | XP_005166762.1 | 3 - 4 | 619 | 81 | ||
| GenBank mRNA | AF362684.1 | - | 7 - 8 | 1171 | 81 | |
| BC092697.1 | - | 7 - 8 | 1340 | 81 | ||
| BC165184.1 | - | 7 - 8 | 1005 | 81 | ||
| JN105111.1 | - | 3 - 4 | 230 | 81 | ||
More Information
Set Membership: |
|

tfe3a
Genomic Map