Assay Details
| Gene Symbol: igf1ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152968.1 | NP_694500.1 | 5 - 6 | 1239 | 87 | |
| XM_017351896.2 | XP_017207385.1 | 5 - 6 | 2171 | 87 | ||
| GenBank mRNA | AF400275.1 | - | 5 - 6 | 1239 | 87 | |
| BC163723.1 | - | 5 - 6 | 1303 | 87 | ||
More Information
Set Membership: |
|

igf1ra
Genomic Map