Assay Details
| Gene Symbol: tnnc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181498.3 | NP_852475.3 | 3 - 4 | 230 | 89 | |
| GenBank mRNA | AF434188.1 | - | 3 - 4 | 196 | 89 | |
| BC083392.1 | - | 230 | 89 | |||
| BC164877.1 | - | 3 - 4 | 230 | 89 | ||
More Information
Set Membership: |
|

tnnc1a
Genomic Map