Assay Details
| Gene Symbol: tnnc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181498.3 | NP_852475.3 | 5 - 6 | 490 | 130 | |
| GenBank mRNA | BC083392.1 | - | 490 | 130 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tnnc1a| Gene Symbol: tnnc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181498.3 | NP_852475.3 | 5 - 6 | 490 | 130 | |
| GenBank mRNA | BC083392.1 | - | 490 | 130 | ||
Set Membership: |
|