Assay Details
| Gene Symbol: serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182863.1 | NP_878283.1 | 6 - 7 | 1215 | 69 | |
| GenBank mRNA | AF515272.1 | - | 6 - 7 | 1215 | 69 | |
| BC081642.1 | - | 6 - 7 | 1177 | 69 | ||
More Information
Set Membership: |
|

serpinc1
Genomic Map