Assay Details
Gene Symbol: keap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182864.2 | NP_878284.2 | 3 - 4 | 1255 | 74 | |
GenBank mRNA | AB081315.2 | - | 3 - 4 | 1191 | 74 | |
BC094989.1 | - | 1223 | 74 | |||
BC117613.1 | - | 3 - 4 | 1255 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
