Assay Details
Gene Symbol: keap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182864.2 | NP_878284.2 | 6 - 7 | 2318 | 70 | |
GenBank mRNA | AB081315.2 | - | 6 - 7 | 2254 | 70 | |
BC117613.1 | - | 6 - 7 | 2318 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
