Assay Details
| Gene Symbol: spi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328368.1 | NP_001315297.1 | 2 - 3 | 452 | 87 | |
| NM_001328369.1 | NP_001315298.1 | 2 - 3 | 449 | 87 | ||
| NM_198062.2 | NP_932328.2 | 3 - 4 | 542 | 87 | ||
| XM_005168848.4 | XP_005168905.1 | 3 - 4 | 590 | 87 | ||
| GenBank mRNA | AF321099.1 | - | 3 - 4 | 211 | 87 | |
| AY293624.1 | - | 3 - 4 | 225 | 87 | ||
| BC056282.1 | - | 2 - 3 | 155 | 87 | ||
| BC067622.1 | - | 3 - 4 | 242 | 87 | ||
More Information
Set Membership: |
|

spi1b
Genomic Map