Assay Details
| Gene Symbol: gnaia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198805.2 | NP_942100.1 | 4 - 5 | 811 | 94 | |
| GenBank mRNA | BC049537.1 | - | 4 - 5 | 774 | 94 | |
| EH537458.1 | - | 4 - 5 | 402 | 94 | ||
More Information
Set Membership: |
|

gnaia
Genomic Map