Assay Details
| Gene Symbol: klhl41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198979.1 | NP_945330.1 | 5 - 6 | 1804 | 65 | |
| GenBank mRNA | BC054912.1 | - | 5 - 6 | 1804 | 65 | |
More Information
Set Membership: |
|

klhl41b
Genomic Map