Assay Details
| Gene Symbol: ahcy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199218.1 | NP_954688.1 | 5 - 6 | 670 | 64 | |
| GenBank mRNA | AY398307.1 | - | 5 - 6 | 670 | 64 | |
| BC044200.1 | - | 5 - 6 | 638 | 64 | ||
| BC164526.1 | - | 5 - 6 | 596 | 64 | ||
More Information
Set Membership: |
|

ahcy
Genomic Map