Assay Details
| Gene Symbol: nt5c3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199560.1 | NP_955854.1 | 6 - 7 | 567 | 61 | |
| GenBank mRNA | BC054650.1 | - | 6 - 7 | 567 | 61 | |
More Information
Set Membership: |
|

nt5c3a
Genomic Map