Assay Details
| Gene Symbol: cops3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199685.1 | NP_955979.1 | 842 | 67 | ||
| GenBank mRNA | BC045415.1 | - | 842 | 67 | ||
| BC064288.1 | - | 822 | 67 | |||
| BC095590.1 | - | 868 | 67 | |||
| BC154523.1 | - | 870 | 67 | |||
| BC165863.1 | - | 792 | 67 | |||
More Information
Set Membership: |
|

cops3
Genomic Map