Assay Details
| Gene Symbol: tnika | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199703.1 | NP_955997.1 | 13 - 14 | 1598 | 103 | |
| XM_005163389.4 | XP_005163446.1 | 13 - 14 | 1629 | 103 | ||
| XM_005163392.4 | XP_005163449.1 | 13 - 14 | 1629 | 103 | ||
| GenBank mRNA | BC055134.1 | - | 13 - 14 | 1598 | 103 | |
More Information
Set Membership: |
|

tnika
Genomic Map